Learn More
Strem, An Ascensus Company (COD)Rh(I)Cl dimer, 98% 250mg
SDP
Catalog No. 5090112146
Encompass
CAS# 12092-47-6. 250mg. Chloro(1,5-cyclooctadiene)rhodium(I) dimer, 98%. MFCD00012415. Molecular Weight: 493.08. Molecular Formula: [RhCl(1,5-C8H12)]2. Color/form: yellow to orange xtl. Strem# 45-0380. http://www.strem.com/catalog/v/45-0380/
Catalog No. 50-901-12146 Supplier Strem, An Ascensus Company Supplier No. 450380250MG
CAS# 12092-47-6. 250mg. Chloro(1,5-cyclooctadiene)rhodium(I) dimer, 98%. MFCD00012415. Molecular Weight: 493.08. Molecular Formula: [RhCl(1,5-C8H12)]2. Color/form: yellow to orange xtl. Strem# 45-0380. http://www.strem.com/catalog/v/45-0380/