Learn More
Strem, An Ascensus Company CAS# 12112-67-3. Chloro-1,5-cyclooctadiene iridium(I) dimer, 99% MFCD00012414. 500MG.
SDP
Catalog No. 5090113108
Encompass
CAS# 12112-67-3. 500MG. Chloro-1,5-cyclooctadiene iridium(I) dimer, 99%. MFCD00012414.Molecular Formula: C16H24Cl2Ir2. Molecular Weight: 671.71. Color/Form: red to orange pwdr. Strem#: 77-0400. www.strem.com/catalog/v3/77-0400/
Catalog No. 50-901-13108 Supplier Strem, An Ascensus Company Supplier No. 770400500MG
CAS# 12112-67-3. 500MG. Chloro-1,5-cyclooctadiene iridium(I) dimer, 99%. MFCD00012414.Molecular Formula: C16H24Cl2Ir2. Molecular Weight: 671.71. Color/Form: red to orange pwdr. Strem#: 77-0400. www.strem.com/catalog/v3/77-0400/