Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloro(1,5-cyclooctadiene)iridium(I) dimer, Ir 57.2%

Catalog No. AA1274906
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1274906 5 g
AA1274901 0.25 g
AA1274903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1274906 Supplier Thermo Scientific Chemicals Supplier No. 01274906
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 12112-67-3 | C16H24Cl2Ir2 | 671.70 g/mol

Chloro(1,5-cyclooctadiene)iridium(I) dimer is widely used as a precursor to other iridium complexes, which finds application in homogeneous catalysis like carbonylation, hydrosilylation, hydrofomylation, asymmetric allylic substitutions, metathesis and chiral catalysis reactions. It is involved in the preparation of Crabtree's catalyst, which is used for hydrogenation and hydrogen-transfer reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12112-67-3
Molecular Formula C16H24Cl2Ir2
Molecular Weight (g/mol) 671.70
MDL Number MFCD00012414
InChI Key ZFOUDQNHNLDNLD-MIXQCLKLSA-L
Synonym chloro 1,5-cyclooctadiene iridium i dimer, bis 1,5-cyclooctadiene diiridium i dichloride, 1z,5z-cycloocta-1,5-diene; iridium; dichloride, iridium, di-.mu.-chlorobis 1,2,5,6-.eta.-1,5-cyclooctadiene di, ir2cl2 cod 2, chloro-1,5-cyclooctadiene iridium i dimer, 1z,5z-cycloocta-1,5-diene; iridium 1+
PubChem CID 6436381
IUPAC Name (1Z,5Z)-cycloocta-1,5-diene;iridium;dichloride
SMILES [Cl-].[Cl-].[Ir+].[Ir+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

Specifications

Melting Point 190°C (decomposition)
Color White
Odor Odorless
Quantity 5 g
Solubility Information Soluble in chloroform and toluene. Slightly soluble in acetone and alcohol. Insoluble in water.
Formula Weight 671.71
Physical Form Crystalline Powder
Chemical Name or Material Chloro(1, 5-cyclooctadiene)iridium(I) dimer
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 235-170-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.