Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloro(1,5-cyclooctadiene)iridium(I) Dimer 93.0+%, TCI America™
SDP

Catalog No. C18071G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18071G 1 g
C1807250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C18071G Supplier TCI America Supplier No. C18071G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 12112-67-3
Molecular Formula C16H24Cl2Ir2
Molecular Weight (g/mol) 671.70
MDL Number MFCD00012414
InChI Key ZFOUDQNHNLDNLD-MIXQCLKLSA-L
Synonym chloro 1,5-cyclooctadiene iridium i dimer, bis 1,5-cyclooctadiene diiridium i dichloride, 1z,5z-cycloocta-1,5-diene; iridium; dichloride, iridium, di-.mu.-chlorobis 1,2,5,6-.eta.-1,5-cyclooctadiene di, ir2cl2 cod 2, chloro-1,5-cyclooctadiene iridium i dimer, 1z,5z-cycloocta-1,5-diene; iridium 1+
PubChem CID 6436381
IUPAC Name diiridium(1+) bis((1Z,5Z)-cycloocta-1,5-diene) dichloride
SMILES [Cl-].[Cl-].[Ir+].[Ir+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1

Specifications

Melting Point 205°C
Quantity 1 g
Formula Weight 671.70
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chloro(1,5-cyclooctadiene)iridium(I) Dimer
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.