Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II)

Catalog No. AA46803MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA46803MD 250 mg
AA4680303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA46803MD Supplier Thermo Scientific Chemicals Supplier No. 046803MD
Add to Cart

CAS: 92390-26-6 | C18H27ClRu | 379.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92390-26-6
Molecular Formula C18H27ClRu
Molecular Weight (g/mol) 379.93
MDL Number MFCD07369035
InChI Key RSZMVRFBKKVJKS-PHFPKPIQSA-M
IUPAC Name Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II)
SMILES Cl[Ru++].C1C\C=C/CC\C=C/1.Cc1c(C)c(C)c(C)c1C

Specifications

Melting Point 143°C to 147°C
Quantity 250 mg
UN Number UN3395
Synonym Pentamethylcyclopentadienyl(1,5-cyclooctadiene)ruthenium(II) chloride; RuCl(cod)Cp
Formula Weight 379.94
Physical Form Crystalline Solid
Chemical Name or Material Chloro(1,5-cyclooctadiene)(pentamethylcyclopentadienyl)ruthenium(II)
Hazard Category H261
Hazard Statement GHS H Statement
H261
In contact with water releases flammable gas.
Precautionary Statement P223-P231+P232-P280-P335+P334-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.3; Packaging Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, SOLID, WATER-REACTIVE
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.