Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Chloro 2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-glucopyranose | 3068-34-6 | 98.0% | 365.76 | 1 G
SDP

Catalog No. 5000452078
Encompass_Preferred

Chloro 2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-glucopyranose is a biochemical reagent utilized in glycobiology research. This field explores the structure, synthesis, biology, and evolution of sugars, encompassing carbohydrate chemistry, the enzymology of glycan formation and degradation, protein-glycan recognition, and the role of glycans within biological systems. It is relevant to basic research, biomedicine, and biotechnology.

  • Molecular weight: 365.76
  • Formula: C14H20ClNO8
  • CAS no.: 3068-34-6
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 98.0%

Catalog No. 50-004-52078 Supplier Medchemexpress LLC Supplier No. HYW0110951G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chloro 2-acetamido-2-deoxy-3,4,6-tri-O-acetyl-α-D-glucopyranose is a biochemical reagent utilized in glycobiology research. This field explores the structure, synthesis, biology, and evolution of sugars, encompassing carbohydrate chemistry, the enzymology of glycan formation and degradation, protein-glycan recognition, and the role of glycans within biological systems. It is relevant to basic research, biomedicine, and biotechnology.

  • Molecular weight: 365.76
  • Formula: C14H20ClNO8
  • CAS no.: 3068-34-6
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 98.0%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.