Learn More
Strem, An Ascensus Company CAS# 1142811-12-8. 500mg. Chloro(2-di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)[2-(2-aminoethyl)phenyl] palladium(II), min. 98% [t-BuXPhos Palladacycle Gen. 1]. MFCD12911909
SDP
Supplier:  Strem, An Ascensus Company 460264500MG
Encompass
CAS# 1142811-12-8. 500mg. Chloro(2-di-t-butylphosphino-2',4',6'-tri-i-propyl-1,1'-biphenyl)[2-(2-aminoethyl)phenyl] palladium(II), min. 98% [t-BuXPhos Palladacycle Gen. 1]. MFCD12911909. Molecular Weight: 686.69. Molecular Formula: C37H55ClNPPd. Color/form: white pwdr. Strem# 46-0264. http://www.strem.com/catalog/v/46-0264/
Catalog No. NC9881485