Learn More
Strem, An Ascensus Company CAS# 1702744-45-3. 500mg. Chloro(2-methylphenyl)(N,N,N',N'-tetramethyl-1,2-ethylenediamine)nickel(II), 99% (contains about 10% o-chlorotoluene) NiCl(o-tolyl)(TMEDA).
SDP
Supplier:  Strem, An Ascensus Company 280165500MG
Encompass
CAS# 1702744-45-3. 500mg. Chloro(2-methylphenyl)(N,N,N',N'-tetramethyl-1,2-ethylenediamine)nickel(II), 99% (contains about 10% o-chlorotoluene) NiCl(o-tolyl)(TMEDA). . Molecular Weight: 301.48. Molecular Formula: C13H23ClN2Ni. Color/form: dark-orange pwdr. Strem# 28-0165. http://www.strem.com/catalog/v/28-0165/
Catalog No. NC1132920