Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloro(4-Cyanophenyl)[™-1-[(S)-2-(Diphenylphosphino)Ferrocenyl]Ethylditertbutylphosphine]Nickel(II)

Catalog No. 900944250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-094-4250MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 90-094-4250MG Supplier Sigma Aldrich Supplier No. 900944250MG
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Amination of aryl chlorides with ammonia.Nickel-Catalyzed Amination of Aryl Chlorides with Ammonia or Ammonium SaltsValorization of the Primary Building Blocks Ammonia and Acetone Featuring Pd- and Ni-Catalyzed Monoarylations

Specifications

CAS 1859379-85-3
Molecular Formula C26H35N3O11
Linear Formula C26H35N3O11
Quantity 50 mg
Synonym 1-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)-3,6,9,12,15-pentaoxaoctadecan-18-oic acid; Crosslinker-E3 Ligase ligand conjugate; Protein degrader building block for PROTAC(R) research; Template for synthesis of targeted protein degrader
Molecular Weight (g/mol) 565.57 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.