Learn More
Strem, An Ascensus Company CAS# 1375325-77-1. 500mg. Chloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene][2'-amino-1,1'-biphenyl]palladium(II) dichloromethane adduct, min. 98% [Xantphos Palladacycle Gen. 2]. MFCD28411853
SDP
Catalog No. 501385536
Encompass
CAS# 1375325-77-1. 500mg. Chloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene][2'-amino-1,1'-biphenyl]palladium(II) dichloromethane adduct, min. 98% [Xantphos Palladacycle Gen. 2]. MFCD28411853. Molecular Weight: 888.71. Molecular Formula: C51H42ClNOP2Pd. Color/form: pale yellow pwdr. Strem# 46-0955. http://www.strem.com/catalog/v/46-0955/
Catalog No. 50-138-5536 Supplier Strem, An Ascensus Company Supplier No. 460955500MG
CAS# 1375325-77-1. 500mg. Chloro[9,9-dimethyl-4,5-bis(diphenylphosphino)xanthene][2'-amino-1,1'-biphenyl]palladium(II) dichloromethane adduct, min. 98% [Xantphos Palladacycle Gen. 2]. MFCD28411853. Molecular Weight: 888.71. Molecular Formula: C51H42ClNOP2Pd. Color/form: pale yellow pwdr. Strem# 46-0955. http://www.strem.com/catalog/v/46-0955/