Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloro(Chloromethyl)Dimethylsilane

Catalog No. 226181100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-618-1100G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 22-618-1100G Supplier Sigma Aldrich Supplier No. 226181100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chloro(Chloromethyl)Dimethylsilane

Specifications

CAS 53531-34-3
Melting Point 132°C to 135°C
Molecular Formula C16H11F3O
Refractive Index n20/D 1.437
Linear Formula C16H11F3O
MDL Number MFCD00001260
Quantity 100 mg
Synonym (Chloromethyl)dimethylchlorosilane; CMDMCS
Molecular Weight (g/mol) 276.25 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.