Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloro(dimethyl)vinylsilane

Catalog No. 39543925ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-543-925ML 25 mL
39-543-9100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 39-543-925ML Supplier Sigma Aldrich Supplier No. 39543925ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Chloro(dimethyl)vinylsilane (dimethylvinylchlorosilane, vinyldimethylchlorosilane, c4h9clsi) is an organosilicon compound
  • It participates in the preparation of 1,1,2,2-tetramethyl-1,2-divinyldisilane
  • It undergoes [2+4] cycloaddition reaction with t-butyllithium in the presence of 2,3-dimethyl-1,3-butadiene to afford cycloadducts.chloro(dimethyl)vinylsilane may be used to prepare silicon-containing polymers, silaheterocycles, and new chelating ligands

Specifications

CAS 1719-58-0
Density 0.874 g/mL (at 25°C (literature))
Boiling Point 82°C to 83°C (lit.)
Molecular Formula C4H9ClSi
Refractive Index n20/D 1.414 (literature)
Linear Formula H2C=CHSi(CH3)2 Cl
Quantity 25 mL
MDL Number MFCD00018090
Synonym Dimethylvinylchlorosilane; Vinyldimethylchlorosilane
Molecular Weight (g/mol) 120.65
Percent Purity 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.