Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloro(Dodecyl)Dimethylsilane

Catalog No. 4024225ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-242-25ML 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 40-242-25ML Supplier Sigma Aldrich Supplier No. 4024225ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chloro(Dodecyl)Dimethylsilane

Specifications

CAS 18437-78-0
Melting Point 79°C to 83°C
Molecular Formula C18H12F3P
Refractive Index n20/D 1.445
Linear Formula (FC6H4)3P
MDL Number MFCD00013553
Quantity 5 g
Synonym Tri-(p-fluorophenyl)phosphine
Molecular Weight (g/mol) 316.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.