Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloro(dodecyl)dimethylsilane 95.0+%, TCI America™
SDP

Catalog No. C146925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C146925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C146925ML Supplier TCI America Supplier No. C146925ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 66604-31-7
Molecular Formula C14H31ClSi
Molecular Weight (g/mol) 262.937
MDL Number MFCD00043326
InChI Key DLLABNOCKQMTEJ-UHFFFAOYSA-N
Synonym dodecyldimethylchlorosilane, chloro dodecyl dimethylsilane, 1-chlorododecyl-dimethyl-silicon, chlorododecyldimethylsilane, acmc-209nvw, dimethyldodecylchlorosilane, dimethyldodecylsilyl chloride, n-dodecyldimethylchlorosilane, dimethyl-dodecyl-chlorosilane, chloro-dimethyl-dodecylsilane
PubChem CID 2758731
IUPAC Name chloro-dodecyl-dimethylsilane
SMILES CCCCCCCCCCCC[Si](C)(C)Cl

Specifications

Color Yellow
Quantity 25 mL
UN Number 2987
Formula Weight 262.94
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Chloro(dodecyl)dimethylsilane
EINECSNumber (2)-2041
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.