Learn More
Medchemexpress LLC HY-107373 100mg Medchemexpress, β-Chloro-L-alanine L-β-Chloroalanine CAS:2731-73-9 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY107373100MG
Encompass_Preferred
Medchemexpress,HY-107373 100mg β-Chloro-L-alanine L-β-Chloroalanine CAS:2731-73-9 Formula:C3H6ClNO2 β-Chloro-L-alanine is a bacteriostatic amino acid analog which inhibits a number of enzymes, including threonine deaminase and alanine racemase. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7332