Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

βChloro-L-Alanine, MP Biomedicals

Catalog No. ICN202067 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN202067 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN202067 Supplier MP Biomedicals, Inc Supplier No. 0520206780

Chemical Identifiers

CAS 2731-73-9
Molecular Formula C3H6ClNO2
Molecular Weight (g/mol) 123.536
InChI Key ASBJGPTTYPEMLP-REOHCLBHSA-N
Synonym 3-chloro-l-alanine, r-2-amino-3-chloropropanoic acid, alanine, 3-chloro, beta-chloro-l-alanine, h-ala cl-oh, 2r-2-amino-3-chloropropanoic acid, l-alanine, 3-chloro, 2r-beta-chloroalanine, alanine, 3-chloro-, l, snxhmvhai jihl@
PubChem CID 439772
ChEBI CHEBI:17403
IUPAC Name (2R)-2-amino-3-chloropropanoic acid
SMILES C(C(C(=O)O)N)Cl

Specifications

Quantity 100 mg
Formula Weight 123.6
Chemical Name or Material β-Chloro-L-Alanine
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.