Learn More
Abcam 2-Chloro-4-nitrophenyl-ß-D-cellobioside, 25MG
Catalog No. 50996129
Encompass_Preferred
MW 497.8 Da, Purity >95%. Chromogenic substrate for cellulase activity. Suitable for kinetic characterisation of cellulases. Useful for biofuels research to measure cellulase activity for optimum ethanol production.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-129 Supplier Abcam Supplier No. AB27521025MG
MW 497.8 Da, Purity >95%. Chromogenic substrate for cellulase activity. Suitable for kinetic characterisation of cellulases. Useful for biofuels research to measure cellulase activity for optimum ethanol production.
The product is subject to the following: Abcam Restricted Use Statement