Learn More
Medchemexpress LLC Chloro-PEG5-chloride 25mg
SDP
Catalog No. 5000206244
Encompass_Preferred
Chloro-PEG5-chloride is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-06244 Supplier Medchemexpress LLC Supplier No. 25MGHYW096131
Chloro-PEG5-chloride is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]