Learn More
Strem, An Ascensus Company CAS# 113860-07-4. 2g. Chloro(pentamethylcyclopentadienyl)ruthenium(II) tetramer, min. 95%. MFCD03792150
SDP
Catalog No. 5090111815
Encompass
CAS# 113860-07-4. 2g. Chloro(pentamethylcyclopentadienyl)ruthenium(II) tetramer, min. 95%. MFCD03792150. Molecular Weight: 1087.01. Molecular Formula: [C10H15RuCl]4. Color/form: dark red xtl. Strem# 44-0145. http://www.strem.com/catalog/v/44-0145/
Catalog No. 50-901-11815 Supplier Strem, An Ascensus Company Supplier No. 4401452G
CAS# 113860-07-4. 2g. Chloro(pentamethylcyclopentadienyl)ruthenium(II) tetramer, min. 95%. MFCD03792150. Molecular Weight: 1087.01. Molecular Formula: [C10H15RuCl]4. Color/form: dark red xtl. Strem# 44-0145. http://www.strem.com/catalog/v/44-0145/