Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloro[(tri-tert-butylphosphine)-2-(2-aminobiphenyl)]palladium(II) 98.0+%, TCI America™
SDP

Catalog No. C27341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C27341G Supplier TCI America Supplier No. C27341G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1375325-71-5
Molecular Formula C24H37ClNPPd
MDL Number MFCD21608496

Specifications

Melting Point 159°C
Color Yellow
Quantity 1 g
Formula Weight 512.41
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chloro[(tri-tert-butylphosphine)-2-(2-aminobiphenyl)]palladium(II)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.