Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloro(Triphenylphosphine)Gold(I)

Catalog No. 254037500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-403-7500MG 500 mg
25-403-75G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 25-403-7500MG Supplier Sigma Aldrich Supplier No. 254037500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent is used with cocatalytic halide abstraction agent to activate pi-systems toward addition by heteroatom and carbon nucleophiles.

Specifications

CAS 14243-64-2
Molecular Formula C18H15AuClP
Linear Formula [(C6H5)3P]AuCl
Quantity 500 mg
MDL Number MFCD00009588
Synonym (Ph3P)AuCl; Triphenylphosphinegold(I) chloride
Molecular Weight (g/mol) 494.71 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.