Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroacetic acid, ACS, 99%

Manufacturer:  Alfa Aesar 4172436

Quantity 500g

 View more versions of this product

Catalog No. AA4172436


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

79-11-8
CH2CO2H
MFCD00002683
605438
Chloroacetic Acid
ClCH2COOH
94.5
UN1751
14,2112
ACS Grade

Specifications

500g
Crystalline
CH2CO2H
MFCD00002683
605438
Chloroacetic Acid
ACS Grade
79-11-8
ClCH2COOH
94.5
UN1751
14,2112
99%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H311-H331-H314

GHS P Statement
P260-P261-P280-P301+P310-P303+P361+P353-P305+P351+P338-P361-P301+P330+P331-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROACETIC ACID, SOLID

RTECSNumber : AF8575000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit