Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroacetic Acid (Certified), Fisher Chemical

Catalog No. A176500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A176-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A176-500

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 79-11-8
Molecular Formula C2H3ClO2
Molecular Weight (g/mol) 94.494
MDL Number MFCD00002683
InChI Key FOCAUTSVDIKZOP-UHFFFAOYSA-N
Synonym chloroacetic acid, monochloroacetic acid, chloracetic acid, acetic acid, chloro, acide chloracetique, chloroethanoic acid, monochloroethanoic acid, monochloracetic acid, monochloorazijnzuur, monochloressigsaeure
PubChem CID 300
ChEBI CHEBI:27869
IUPAC Name 2-chloroacetic acid
SMILES C(C(=O)O)Cl

Specifications

Melting Point 61°C
Color White
Density 1.580g/cm³
pH 1.90
Boiling Point 189°C
Melting Range ≥61.3°C to ≤63.3°C
Packaging Glass Bottle
Linear Formula CH2ClCOOH
Quantity 500 g
UN Number 1751
Vapor Density 3.2g/mol
Formula Weight 94.5
Grade Certified
Physical Form Solid
Chemical Name or Material Chloroacetic Acid
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation Class 6.1: Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.