Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroacetone, 96%, stabilized

Catalog No. AC173470050
Click to view available options
Quantity:
25 mL
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum

Catalog No. AC173470050

Supplier: Thermo Scientific Chemicals 173470050

Only null left
Add to Cart

CAS: 78-95-5 | C3H5ClO | 92.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-95-5
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
MDL Number MFCD00000936
InChI Key BULLHNJGPPOUOX-UHFFFAOYSA-N
Synonym chloroacetone, chloropropanone, acetonyl chloride, monochloroacetone, tonite, 2-propanone, 1-chloro, chloro-2-propanone, 1-chloroacetone, chloromethyl methyl ketone, 1-chloro-2-propanone
PubChem CID 6571
ChEBI CHEBI:47220
IUPAC Name 1-chloropropan-2-one
SMILES CC(=O)CCl

Specifications

Melting Point -44.5°C
Color Brown-Yellow to Yellow
Density 1.1600g/mL
Boiling Point 119.0°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Plastic drum
Refractive Index 1.4310 to 1.4330
Linear Formula ClCH2COCH3
Quantity 5 L
Beilstein 01, 653
Specific Gravity 1.16
Merck Index 15, 2114
Solubility Information Solubility in water: 124g/L (20°C). Other solubilities: soluble in alcohol, ether and chloroform
Stabilizer 0.1% epoxidized soybean oil
Viscosity 1.11 mPa.s (25°C)
Formula Weight 92.52
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Chloroacetone, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Fatal if inhaled.
Causes ski
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
EINECSNumber 201-161-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.