Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chloroacetyl chloride

Catalog No. 22880500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-880-500ML 500 mL
22-880-100ML 100 mL
22-880-1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-880-500ML Supplier Sigma Aldrich Supplier No. 22880500ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chloroacetyl chloride

Specifications

Boiling Point 105°C to 106°C (lit.)
Melting Point -22°C (lit.)
CAS 79-04-9
Quantity 500 mL
Refractive Index n20/D 1.453 (literature); n20/D 1.453
Density 1.419 g/mL at 20°C, 1.418 g/mL (at 25°C (literature))
Molecular Formula C2H2Cl2O
Linear Formula ClCH2COCl
MDL Number MFCD00000725
Molecular Weight (g/mol) 112.94
Percent Purity ≥99.0% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.