Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroacetyl chloride, 98%

Catalog No. AAA15846AP
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA15846AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA15846AP Supplier Thermo Scientific Chemicals Supplier No. A15846AP

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

missing translation for 'validMainframeMsg'

CAS: 79-04-9 | C2H2Cl2O | 112.937 g/mol

Chloroacetyl chloride is an intermediate in the production of alachlor and butachlor. It is used to prepare phenacyl chloride through Friedel-Crafts acylation of benzene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetyl chloride is an intermediate in the production of alachlor and butachlor. It is used to prepare phenacyl chloride through Friedel-Crafts acylation of benzene.

Solubility
Miscible with ethyl ether, acetone, benzene and carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents, strong bases, alcohols and water. Moisture sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-04-9
Molecular Formula C2H2Cl2O
Molecular Weight (g/mol) 112.937
MDL Number MFCD00000725
InChI Key VGCXGMAHQTYDJK-UHFFFAOYSA-N
Synonym chloroacetyl chloride, acetyl chloride, chloro, chloracetyl chloride, chloroacetic chloride, monochloroacetyl chloride, chloroacetic acid chloride, chloroacetylchloride, chlorure de chloracetyle, unii-k5uml06yuo, chlorid kyseliny chloroctove
PubChem CID 6577
ChEBI CHEBI:34624
IUPAC Name 2-chloroacetyl chloride
SMILES C(C(=O)Cl)Cl

Specifications

Melting Point -22°C
Density 1.417
Boiling Point 105°C to 106°C
Odor Pungent
Refractive Index 1.453
Linear Formula ClCH2COCl
Quantity 500 mL
UN Number UN1752
Beilstein 605439
Sensitivity Moisture sensitive
Merck Index 14,2067
Solubility Information Miscible with ethyl ether,acetone,benzene and carbon tetrachloride.
Formula Weight 112.94
Percent Purity 98%
Chemical Name or Material Chloroacetyl chloride
Show More Show Less
Hazard Category H290-H301+H311+H331-H314-H335-H372
Hazard Statement GHS H Statement
H301-H311-H331-H372-H314
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Precautionary Statement P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: CHLOROACETYL CHLORIDE
EINECSNumber 201-171-6
RTECSNumber AO6475000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.