Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroacetyl chloride, 98%

Catalog No. AC147292500
Change view
Click to view available options
Quantity:
100 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC147292500 250 mL Glass bottle
AC147291000 100 mL Glass bottle
AC147290010 1 L Glass bottle
AC147290025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC147292500 Supplier Thermo Scientific Chemicals Supplier No. 147292500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

CAS: 79-04-9 | C2H2Cl2O | 112.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-04-9
InChI Key VGCXGMAHQTYDJK-UHFFFAOYSA-N
Synonym chloroacetyl chloride, acetyl chloride, chloro, chloracetyl chloride, chloroacetic chloride, monochloroacetyl chloride, chloroacetic acid chloride, chloroacetylchloride, chlorure de chloracetyle, unii-k5uml06yuo, chlorid kyseliny chloroctove
PubChem CID 6577
ChEBI CHEBI:34624
IUPAC Name 2-chloroacetyl chloride
SMILES C(C(=O)Cl)Cl

Specifications

Melting Point -22.0°C
Color Colorless to Yellow
Density 1.4200g/mL
Boiling Point 105.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C2H2Cl2O
Refractive Index 1.4520 to 1.4540
Linear Formula ClCH2COCl
Quantity 250 mL
Beilstein 02,199
Fieser 01,130; 03,46
Specific Gravity 1.42
Merck Index 15,2067
Solubility Information Solubility in water: reacts. Other solubilities: miscible in ethyl ether, soluble in acetone, carbontetrachloride
Molecular Weight (g/mol) 112.94
Formula Weight 112.94
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Chloroacetyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if inhaled.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Very toxic to aquatic life.<br
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
EINECSNumber 201-171-6

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.