Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroacetylglycylglycine 98.0+%, TCI America™
SDP

Catalog No. C009925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C009925G 25 g
C00991G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C009925G Supplier TCI America Supplier No. C009925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 15474-96-1 | C6H9ClN2O4 | 208.60 g/mol

Chemical Identifiers

CAS 15474-96-1
Molecular Formula C6H9ClN2O4
Molecular Weight (g/mol) 208.60
MDL Number MFCD00055928
InChI Key NCOXSGKCQMMTRX-UHFFFAOYSA-N
PubChem CID 259611
IUPAC Name 2-[2-(2-chloroacetamido)acetamido]acetic acid
SMILES OC(=O)CNC(=O)CNC(=O)CCl

Specifications

Melting Point 179°C
Color White
Packaging Glass Bottle with Plastic Insert
Quantity 25 g
Formula Weight 208.60
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chloroacetylglycylglycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.