Learn More
ATCC Mycobacterium mucogenicum; Strain MO76
Supplier:  ATCC 49650
Encompass
Mycobacterium mucogenicum strain MO76 is a bacterial type strain that was isolated in Texas from an infected thyroglossal duct cyst. This strain is propagated aerobically at 37°C.
Catalog No. 50-238-1229