Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorobenzene, ≥99.7% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019795 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9795 5 mL
11-101-8369 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9795 Supplier MilliporeSigma Supelco Supplier No. 086505MLF
Add to Cart
Add to Cart
Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 108-90-7
Ignition Point 1178°F
Vapor Pressure 11.8 mmHg (25°C)
Vapor Density 3.86 (vs. air)
Density 1.106 g/mL (at 25°C)
Flash Point 27.0°C - closed cup
Linear Formula C6H5 Cl
MDL Number MFCD00000530
Melting Point -45°C
Boiling Point 132°C
Type Analytical Standard
Quantity 5 mL
Formula Weight 112.56
Percent Purity ≥99.7% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.