Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorobenzene, 99.9%, for HPLC

Catalog No. AC445650010
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC445650010 1 L Glass bottle
AC445651000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC445650010 Supplier Thermo Scientific Chemicals Supplier No. 445650010
Get 1 Extra Chemical When You Buy 2
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 108-90-7 | C6H5Cl | 112.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.56
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
Synonym monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
PubChem CID 7964
ChEBI CHEBI:28097
IUPAC Name chlorobenzene
SMILES C1=CC=C(C=C1)Cl

Specifications

Melting Point -45.0°C
Density 1.1075g/mL
Quantity 1 L
Packaging Glass bottle
Refractive Index 1.5230 to 1.5250
Specific Gravity 1.1075
Assay Percent Range 99.85% min. (GC)
Linear Formula C6H5Cl
Viscosity 0.8 mPa.s (20°C)
Merck Index 15,2122
Boiling Point 131.0°C
Solubility Information Solubility in water: 0.4g/L (20°C). Other solubilities: miscible with most common organic solvents
Formula Weight 112.56
Percent Purity 99.9%
Grade HPLC
Chemical Name or Material Chlorobenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly
EINECSNumber 203-628-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.