Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorobenzene/Acetic Acid, 67/33 v/v, Reagents

Catalog No. 01335659
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-659 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-659 Supplier Reagents Holdings Llc Supplier No. CC1030004C1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chlorobenzene/Acetic Acid, 67/33 v/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 108-90-7 , 64-19-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.56
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
IUPAC Name chlorobenzene
SMILES ClC1=CC=CC=C1

Specifications

pH 2
Color Colorless
Quantity 4 L
Packaging Safety Coated Glass
Physical Form Liquid
Specific Gravity 1.0879
Grade Reagent
Chemical Name or Material Chlorobenzene/Acetic Acid, 67/33 v/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.