Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorobenzene Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019491 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9491 3 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9491 Supplier MilliporeSigma Supelco Supplier No. PHR16653X1.2ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical secondary standard and certified reference material for quality control analysis.

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chlorobenzene
Packaging Ampule
CAS 108-90-7
Ignition Point 1178°F
Vapor Pressure 11.8 mmHg (25°C)
Vapor Density 3.86 (vs. air)
Density 1.106 g/mL (at 25°C)
Linear Formula C6H5 Cl
Technique HPLC
MDL Number MFCD00000530
Certifications/Compliance ISO 17034
Melting Point -45°C
Boiling Point 132°C
Synonym Chlorobenzene
Type Certified Reference Material
Solvent or Matrix Neat
Industry Type Pharmaceutical
Molecular Weight (g/mol) 112.56
Quantity 3 x 1.2 mL
Formula Weight 112.56
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.