Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorobenzene, puriss. p.a., ≥99.5% (GC), ACS Reagent, Solstice

Catalog No. 60045434 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-434 1 L Glass Bottle
60-045-435 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-045-434 Supplier Solstice Supplier No. 235701L
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent,≥99.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-90-7
Molecular Formula C6H5Cl
Molecular Weight (g/mol) 112.556
MDL Number MFCD00000530
InChI Key MVPPADPHJFYWMZ-UHFFFAOYSA-N
Synonym monochlorobenzene, benzene chloride, phenyl chloride, benzene, chloro, chlorbenzene, chlorobenzol, monochlorbenzol, chlorbenzol, chlorobenzen, clorobenzene
PubChem CID 7964
ChEBI CHEBI:28097
IUPAC Name chlorobenzene
SMILES C1=CC=C(C=C1)Cl

Specifications

Melting Point -45°C
Density 1.106g/mL at 25°C
Quantity 1 L
Explosion Limit 0.071
Flash Point 27°C (80.6°F)
Ignition Point 1178°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.524(lit.)
Vapor Pressure 11.8mmHg (25°C)
UN Number UN1134
Beilstein 605632
Boiling Point 132°C
Formula Weight 112.56g/mol
Percent Purity ≥99.5% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Chlorobenzene
Show More Show Less
Hazard Statement P210-P261-P370 + P378
Precautionary Statement H226-H315-H332-H411
DOTInformation Class 3 - PG 3 - Chlorobenzene
EINECSNumber 203-628-5
RTECSNumber CZ0175000
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.