Learn More
Strem, An Ascensus Company CAS# 12246-51-4. 250mg. Chlorobis(cyclooctene)iridium(I) dimer, 97%. MFCD00213465
SDP
Catalog No. 5090113104
Encompass
CAS# 12246-51-4. 250mg. Chlorobis(cyclooctene)iridium(I) dimer, 97%. MFCD00213465. Molecular Weight: 896.13. Molecular Formula: [IrCl(C8H14)2]2. Color/form: yellow to orange solid Strem# 77-0250. http://www.strem.com/catalog/v/77-0250/
Catalog No. 50-901-13104 Supplier Strem, An Ascensus Company Supplier No. 770250250MG
CAS# 12246-51-4. 250mg. Chlorobis(cyclooctene)iridium(I) dimer, 97%. MFCD00213465. Molecular Weight: 896.13. Molecular Formula: [IrCl(C8H14)2]2. Color/form: yellow to orange solid Strem# 77-0250. http://www.strem.com/catalog/v/77-0250/