Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorobis(cyclooctene)iridium(I) Dimer 97.0+%, TCI America™
SDP

Catalog No. C2985200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2985200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C2985200MG Supplier TCI America Supplier No. C2985200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 12246-51-4
Molecular Formula C32H56Cl2Ir2-2
Molecular Weight (g/mol) 896.134
MDL Number MFCD00213465
InChI Key CJJIQMGSHWWMCK-XFCUKONHSA-L
Synonym chlorobis cyclooctene iridium i dimer, di-mu-chlorotetrakis cyclooctene diiridium i, bis cyclooctene iridium i chloride, dimer, chlorobis cyclooctene-iridium i dimer, chlorobis cyclooctene iridium dimer, di-mu-chlorobis cyclooctene iridium i
PubChem CID 91972094
IUPAC Name cyclooctene;iridium;dichloride
SMILES C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1.C1CCCC=CCC1.[Cl-].[Cl-].[Ir].[Ir]

Specifications

Melting Point 153°C
Quantity 200 mg
Formula Weight 896.13
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Chlorobis(cyclooctene)iridium(I) Dimer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.