Learn More
Strem, An Ascensus Company CAS# 12279-09-3. 1g. Chlorobis(cyclooctene)rhodium(I) dimer, min. 98%. MFCD00013287
SDP
Catalog No. 5090112140
Encompass
CAS# 12279-09-3. 1g. Chlorobis(cyclooctene)rhodium(I) dimer, min. 98%. MFCD00013287. Molecular Weight: 717.50. Molecular Formula: [RhCl(C8H14)2]2. Color/form: orange to red xtl. Strem# 45-0265. http://www.strem.com/catalog/v/45-0265/
Catalog No. 50-901-12140 Supplier Strem, An Ascensus Company Supplier No. 4502651G
CAS# 12279-09-3. 1g. Chlorobis(cyclooctene)rhodium(I) dimer, min. 98%. MFCD00013287. Molecular Weight: 717.50. Molecular Formula: [RhCl(C8H14)2]2. Color/form: orange to red xtl. Strem# 45-0265. http://www.strem.com/catalog/v/45-0265/