Learn More
Strem, An Ascensus Company CAS# 12081-16-2. 250mg. Chlorobis(ethylene)rhodium(I) dimer, 99%. MFCD00013206
SDP
Catalog No. 5090112141
Encompass
CAS# 12081-16-2. 250mg. Chlorobis(ethylene)rhodium(I) dimer, 99%. MFCD00013206. Molecular Weight: 388.93. Molecular Formula: [RhCl(C2H4)2]2. Color/form: rust-colored pwdr. Strem# 45-0270. http://www.strem.com/catalog/v/45-0270/
Catalog No. 50-901-12141 Supplier Strem, An Ascensus Company Supplier No. 450270250MG
CAS# 12081-16-2. 250mg. Chlorobis(ethylene)rhodium(I) dimer, 99%. MFCD00013206. Molecular Weight: 388.93. Molecular Formula: [RhCl(C2H4)2]2. Color/form: rust-colored pwdr. Strem# 45-0270. http://www.strem.com/catalog/v/45-0270/