Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chlorobis(Trimethylsilyl)Methane

Catalog No. 3617475G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-174-75G 5 g
36-174-725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 36-174-75G Supplier Sigma Aldrich Supplier No. 3617475G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chlorobis(trimethylsilyl)methane can be used as a reagent for the preparation of: Para-bis(trimethylsilyl)ethylstyrene (PBTES) monomer, which is used to synthesize corresponding network polymer of styrene. Bis(trimethylsilyl)methyl magnesium chloride (Grignard reagent), which is used in the synthesis of bis(trimethylsilyl) allyl compounds by reacting with alkenyl bromide via the Kumada coupling reaction. N-[Bis(trimethylsilyl)methyl]heterocumulenes.

Specifications

CAS 5926-35-2
Density 0.892 g/mL at 25°C
Boiling Point 57°C to 60°C (15 mmHg, lit.)
Molecular Formula C7H19ClSi2
Refractive Index n20/D 1.449
Linear Formula [(CH3)3Si]2CHCl
MDL Number MFCD00054866
Quantity 5 g
Synonym Bis(trimethylsilyl)chloromethane
Molecular Weight (g/mol) 194.85 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.