Learn More
Strem, An Ascensus Company CAS# 50960-82-2. 1g. Chlorocarbonylgold(I), min. 97%. MFCD00058864
SDP
Supplier:  Strem, An Ascensus Company 7903501G
Encompass
CAS# 50960-82-2. 1g. Chlorocarbonylgold(I), min. 97%. MFCD00058864. Molecular Weight: 260.43. Molecular Formula: Au(CO)Cl. Color/form: off-white pwdr. Strem# 79-0350. http://www.strem.com/catalog/v/79-0350/
Catalog No. 50-901-13387