Learn More
Strem, An Ascensus Company CAS# 1101230-25-4. 100mg. Chlorocarbonylhydrido[4,5-bis-(di-i-propylphosphinomethyl)acridine]ruthenium(II), min.98% Milstein Acridine Catalyst. MFCD19443487
SDP
Supplier:  Strem, An Ascensus Company 440525100MG
Encompass
CAS# 1101230-25-4. 100mg. Chlorocarbonylhydrido[4,5-bis-(di-i-propylphosphinomethyl)acridine]ruthenium(II), min.98% Milstein Acridine Catalyst. MFCD19443487. Molecular Weight: 605.09. Molecular Formula: C28H40ClNOP2Ru. Color/form: orange solid Strem# 44-0525. http://www.strem.com/catalog/v/44-0525/
Catalog No. 50-372-4