Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorocarbonylsulfenyl chloride, 97%

Catalog No. AAL0643206
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0643206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0643206 Supplier Thermo Scientific Chemicals Supplier No. L0643206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2757-23-5 | CCl2OS | 130.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2757-23-5
Molecular Formula CCl2OS
Molecular Weight (g/mol) 130.97
MDL Number MFCD00000703
InChI Key MNOALXGAYUJNKX-UHFFFAOYSA-N
Synonym chlorocarbonylsulfenyl chloride, chlorothio formyl chloride, chloroformylsulfenyl chloride, chloro chlorosulfanyl methanone, chlorothioformyl chloride, chlorocarbonylsulphenyl chloride, chlorocarbonyl sulfenyl chloride, chlorocarbonyl sulphenyl chloride, carbonochloridothioic acid, anhydrosulfide with thiohypochlorous acid, methane, chloro chlorothio oxo
PubChem CID 75990
IUPAC Name S-chloro chloromethanethioate
SMILES ClSC(Cl)=O

Specifications

Density 1.58
Boiling Point 98°C to 101°C
Refractive Index 1.517
Quantity 5 g
UN Number UN3265
Beilstein 506318
Sensitivity Moisture sensitive
Formula Weight 130.98
Percent Purity 97%
Chemical Name or Material Chlorocarbonylsulfenyl chloride
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 220-415-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.