Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorodi(o-tolyl)phosphine, 98%

Catalog No. AAH2589006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2589006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2589006 Supplier Thermo Scientific Chemicals Supplier No. H2589006

missing translation for 'validMainframeMsg'

CAS: 36042-94-1 | C14H14ClP | 248.69 g/mol

Chlorodi(o-tolyl)phosphine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodi(o-tolyl)phosphine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

Solubility
Not miscible or difficult to mix in water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36042-94-1
Molecular Formula C14H14ClP
Molecular Weight (g/mol) 248.69
MDL Number MFCD04038733
InChI Key KAAGXBGJRWFWPT-UHFFFAOYSA-N
Synonym chlorodi o-tolyl phosphine, di-o-tolylchlorophosphine, bis o-tolyl chlorophosphine, chlorobis o-tolyl phosphine, chlorobis 2-methylphenyl phosphane, chloro-bis 2-methylphenyl phosphane, chloroditolylphosphine, chlorodi-o-tolylphosphine, di o-tolyl chlorophosphine, chlorobis 2-methylphenyl phosphine
PubChem CID 4590860
IUPAC Name chloro-bis(2-methylphenyl)phosphane
SMILES CC1=CC=CC=C1P(C2=CC=CC=C2C)Cl

Specifications

Melting Point 57°C
Boiling Point 174°C to 178°C (3 mmHg)
Quantity 5 g
UN Number UN1760
Sensitivity Air and moisture sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 248.69
Percent Purity 98%
Chemical Name or Material Chlorodi(o-tolyl)phosphine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.