Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorodi(p-tolyl)phosphine, 95%

Catalog No. AAH2588906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2588906 5 g
AAH2588903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2588906 Supplier Thermo Scientific Chemicals Supplier No. H2588906
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1019-71-2 | C14H14ClP | 248.69 g/mol

Chlorodi(p-tolyl)phosphine is used as a reactant for synthesis of palladium catalysts for C-C bond forming cross-coupling reactions, ligands for nickel(0)-catalyzed isomerization reactions, phosphinosulfonamide nickel complexes for oligomerization reactions. It is a reactant for phosphinylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorodi(p-tolyl)phosphine is used as a reactant for synthesis of palladium catalysts for C-C bond forming cross-coupling reactions, ligands for nickel(0)-catalyzed isomerization reactions, phosphinosulfonamide nickel complexes for oligomerization reactions. It is a reactant for phosphinylation reactions.

Solubility
Reacts with water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1019-71-2
Molecular Formula C14H14ClP
Molecular Weight (g/mol) 248.69
MDL Number MFCD01630844
InChI Key BJBXRRHIBSXGLF-UHFFFAOYSA-N
Synonym bis 4-methylphenyl chlorophosphine, chlorodi p-tolyl phosphine, chlorodi-p-tolylphosphine, chlorobis 4-methylphenyl phosphine, bis 4-methylphenyl phosphinous chloride, chlorobis 4-methylphenyl phosphane, chloro-bis 4-methylphenyl phosphane, di-p-tolylchlorophosphine, chloro bis-p-tolyl phosphane, di p-tolyl phosphinous chloride
PubChem CID 4188608
IUPAC Name chloro-bis(4-methylphenyl)phosphane
SMILES CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)Cl

Specifications

Density 1.159
Boiling Point 180°C to 184°C (10 mmHg)
Quantity 5 g
UN Number UN1760
Sensitivity Air and moisture sensitive
Solubility Information Reacts with water.
Formula Weight 248.69
Percent Purity 95%
Chemical Name or Material Chlorodi(p-tolyl)phosphine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.