Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorodi(p-tolyl)phosphine 98.0+%, TCI America™
SDP

Catalog No. C32015G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C32015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C32015G Supplier TCI America Supplier No. C32015G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1019-71-2
Molecular Formula C14H14ClP
Molecular Weight (g/mol) 248.69
MDL Number MFCD01630844
InChI Key BJBXRRHIBSXGLF-UHFFFAOYSA-N
Synonym bis 4-methylphenyl chlorophosphine, chlorodi p-tolyl phosphine, chlorodi-p-tolylphosphine, chlorobis 4-methylphenyl phosphine, bis 4-methylphenyl phosphinous chloride, chlorobis 4-methylphenyl phosphane, chloro-bis 4-methylphenyl phosphane, di-p-tolylchlorophosphine, chloro bis-p-tolyl phosphane, di p-tolyl phosphinous chloride
PubChem CID 4188608
IUPAC Name chloro-bis(4-methylphenyl)phosphane
SMILES CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)Cl

Specifications

Color Yellow
Boiling Point 350°C
Quantity 5 g
UN Number 3265
Formula Weight 248.69
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Chlorodi(p-tolyl)phosphine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.