Learn More
Strem, An Ascensus Company CAS# 14523-22-9. 2g. Chlorodicarbonylrhodium(I) dimer. MFCD00135610
SDP
Catalog No. 5090112154
Encompass
CAS# 14523-22-9. 2g. Chlorodicarbonylrhodium(I) dimer. MFCD00135610. Molecular Weight: 388.76. Molecular Formula: [RhCl(CO)2]2. Color/form: red xtl. Strem# 45-0450. http://www.strem.com/catalog/v/45-0450/
Catalog No. 50-901-12154 Supplier Strem, An Ascensus Company Supplier No. 4504502G
CAS# 14523-22-9. 2g. Chlorodicarbonylrhodium(I) dimer. MFCD00135610. Molecular Weight: 388.76. Molecular Formula: [RhCl(CO)2]2. Color/form: red xtl. Strem# 45-0450. http://www.strem.com/catalog/v/45-0450/