Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chlorodicyclohexylborane Solution

Catalog No. 41112450ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-112-450ML 50 mL
41-112-4100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 41-112-450ML Supplier Sigma Aldrich Supplier No. 41112450ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Aldol addition reactionsMukaiyama aldol additionMicrowave assisted ring closing metathesis reactionsC-alkylation of aromatic aldiminesReversible ketone-ketone aldol reactionsSyn- and anti-stereoselective aldol reaction hexenone with aldehydes

Specifications

CAS 36140-19-9
Density 0.752 g/mL at 25°C
Boiling Point 68°C to 69°C
Molecular Formula C12H22BCl
Linear Formula (C6H11)2BCl
MDL Number MFCD00191906
Quantity 50 mL
Synonym Dicyclohexylboron chloride; Dicyclohexylboryl chloride; Dicyclohexylchloroborane
Molecular Weight (g/mol) 212.57 g/mol
Biological Activity Central Nervous System, Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.