Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Chlorodicyclohexylphosphine

Catalog No. 4814081G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-140-81G 1 g
48-140-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 48-140-81G Supplier Sigma Aldrich Supplier No. 4814081G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Chlorodicyclohexylphosphine can be used as a reactant in the synthesis of: 1,2-Bis(dicyclohexylphosphinoxy)ethane ligand by reacting with ethylene glycol in the presence of triethylamine via Michaelis-Arbuzov type rearrangements.1,1,2,2-tetracyclohexyldiphosphine monosulfide ligand by treating with LiS.It can be also used as a starting material for the preparation of some other ligands such as dicyclohexylphosphine oxide , phosphino substituted N-aryl pyrroles , di-tert-butyl((dicyclohexylphosphino)methyl)phosphine , dicyclohexylcyclopentylphosphine. These ligands are used in Pd-catalyzed cross-coupling reactions.

Specifications

CAS 16523-54-9
Density 1.054 g/mL at 25°C
Boiling Point 165°C (12 mmHg, lit.)
Molecular Formula C12H22ClP
Refractive Index n20/D 1.533
Linear Formula (C6H11)2PCl
MDL Number MFCD00014294
Quantity 1 g
Synonym Dicyclohexylchlorophosphine; Dicyclohexylphosphine chloride; Dicyclohexylphosphinous chloride
Molecular Weight (g/mol) 232.73 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.