Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorodifluoroacetic acid, 98+%, Thermo Scientific™

Catalog No. AAA1436922
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1436922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1436922 Supplier Thermo Scientific Chemicals Supplier No. A1436922

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a starting material for generating difluorocarbene. It is also used as a pharmaceutical and agrochemicals intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, bases, active metals and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-04-0
Molecular Formula C2HClF2O2
Molecular Weight (g/mol) 130.475
MDL Number MFCD00004176
InChI Key OAWAZQITIZDJRB-UHFFFAOYSA-N
Synonym chlorodifluoroacetic acid, acetic acid, chlorodifluoro, difluorochloroacetic acid, chloro difluoro acetic acid, 2-chloro-2,2-difluoro-acetic acid, acetic acid, 2-chloro-2,2-difluoro, snpljqdjptadtaetp@, chlorodifluoroaceticacid, chlorodifluoro acetic acid, acmc-209p1k
PubChem CID 66154
IUPAC Name 2-chloro-2,2-difluoroacetic acid
SMILES C(=O)(C(F)(F)Cl)O

Specifications

Melting Point 20°C to 24°C
Density 1.55
Boiling Point 121°C to 122°C
Odor Acrid
Refractive Index 1.3545
Quantity 100 g
UN Number UN3265
Beilstein 956625
Solubility Information Soluble in water.
Formula Weight 130.48
Percent Purity ≥98%
Chemical Name or Material Chlorodifluoroacetic acid
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 200-928-8
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.