Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorodifluoroacetic Acid 98.0+%, TCI America™
SDP

Catalog No. C0990500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0990500G 500 g
C099025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0990500G Supplier TCI America Supplier No. C0990500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 76-04-0
Molecular Formula C2HClF2O2
Molecular Weight (g/mol) 130.475
MDL Number MFCD00004176
InChI Key OAWAZQITIZDJRB-UHFFFAOYSA-N
Synonym chlorodifluoroacetic acid, acetic acid, chlorodifluoro, difluorochloroacetic acid, chloro difluoro acetic acid, 2-chloro-2,2-difluoro-acetic acid, acetic acid, 2-chloro-2,2-difluoro, snpljqdjptadtaetp@, chlorodifluoroaceticacid, chlorodifluoro acetic acid, acmc-209p1k
PubChem CID 66154
IUPAC Name 2-chloro-2,2-difluoroacetic acid
SMILES C(=O)(C(F)(F)Cl)O

Specifications

Boiling Point 126°C
Quantity 500 g
UN Number 3261
Formula Weight 130.47
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Chlorodifluoroacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.